Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Fascaplysin, Synthetic
Click to view available options
Quantity:
1 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | Brown |
Description | 98% by NMR |
Target Proteins | CDK4/D1 |
CAS | 114719-57-2 |
Molecular Formula | C18H11ClN2O |
Formula Weight | 306.8g/mol |
Solubility | DMSO or MeOH |
Content And Storage | Hygroscopic, Protect from light |
Form | Solid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction