Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Gö 6976

Catalog No. 3652501MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
500 μg

Catalog No. 36-525-01MG

Supplier: MilliporeSigma™ 3652501MG

Only null left
Add to Cart
Add to Cart

Gö 6976, CAS 136194-77-9, is a cell-permeable, reversible, and ATP-competitive inhibitor of PKC (IC50 = 7.9 nM for rat brain). Exhibits selectively for PKCα (IC50 = 2.3 nM) and βI (IC50 = 6.2 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins PKC
CAS 136194-77-9
Molecular Formula C24H18N4O
Formula Weight 378.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.