Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Gliotoxin, Gladiocladium fimbriatum
Click to view available options
Quantity:
1 mg
Description
An immunosuppressive secondary metabolite produced by several pathogenic fungi. Immunosuppressive effects are caused by blocking of membrane thiol groups. Causes apoptotic cell death in a variety of cell types including macrophages and thymocytes. Farnesyltransferase (FTase) inhibitor (IC50 = 1.1μM). Reported to increase ryanodine Ca2+ channel activity possibly by oxidizing cysteine residues located on the ryanodine receptor. Specifically inhibits NF-κB activation in B and T cells at nanomolar concentrations.

Specifications
Specifications
Color | White |
Description | 98% by TLC |
Target Proteins | FTase |
CAS | 67-99-2 |
Molecular Formula | C13H14N2O4S2 |
Formula Weight | 326.4g/mol |
Solubility | DMSO or EtOH |
Form | Solid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction