Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ GTP-14564
Click to view available options
Quantity:
5 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | Pale Yellow |
Description | 98% by HPLC |
Target Proteins | Class III receptor tyrosine kinases |
CAS | 34823-86-4 |
Molecular Formula | C15H10N2O |
Formula Weight | 234.3g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction