Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ HMTase Inhibitor V, UNC0224
Click to view available options
Quantity:
5 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Description | 97% by HPLC |
CAS | 1197196-48-7 |
Molecular Formula | C26H43N7O2·H2O |
Formula Weight | 503.7g/mol |
Solubility | DMSO |
Content And Storage | Hygroscopic, Protect from light |
Form | Liquid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction