Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ IKK Inhibitor II, Wedelolactone
Click to view available options
Quantity:
1 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | Yellowish Green |
Description | 95% by HPLC |
Target Proteins | IKKα and β kinase activity |
CAS | 524-12-9 |
Molecular Formula | C16H10O7 |
Formula Weight | 314.3g/mol |
Solubility | DMSO or MeOH |
Content And Storage | Protect from light |
Form | Crystalline Solid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction