Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set III, Lyophilized

Catalog No. p-7060126 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Set
1 mL

The Phosphatase Inhibitor Cocktail Set III, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.