Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ HIF-1 Inhibitor
Click to view available options
Quantity:
5 mg
Description
A cell-permeable amidophenolic compound that inhibits hypoxia-induced HIF-1 transcription activity (IC50 = 0.7 and 2.6μM using AGS and Hep3B cells, respectively, in a reporter assay) by selectively blocking the hypoxia-induced accumulation of cellular HIF-1α protein, while exhibiting no apparent effect on the cellular level of HIF-1α mRNA or that of HIF-1β protein.

Specifications
Specifications
Formulation | A 25 mM (5mg/459μL) solution of HIF-1 Inhibitor in DMSO. |
Description | 95% by HPLC |
Target Proteins | HIF-1 |
CAS | 934593-90-5 |
Molecular Formula | C26H29NO5 |
Formula Weight | 435.5g/mol |
Content And Storage | Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light |
Form | Liquid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction