Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ PP1 Analog
Click to view available options
Quantity:
1 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Formulation | A 10 mM (1mg/315μl) solution of PP1 Analog in DMSO. |
Description | 97% by HPLC |
CAS | 221243-82-9 |
Molecular Formula | C19H19N5 |
Formula Weight | 317.4g/mol |
Content And Storage | Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light |
Form | Liquid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction