Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ JHDM Inhibitor VII, JIB-04
Click to view available options
Quantity:
10 mg
Description
The E-isomer of a cell-permeable pyridine hydrazone compound that acts as a selective inhibitor against Jumonji family histone demethylases (IC50 = 230, 290-320, 340, 435, 445, 855, and 1,100 nM, respectively, against JARID1A/KDM5A, JMJD2D/KDM4D, JMJD2E/KDM4E, JMJD2B/KDM4B, JMJD2A/KMD4A, JMJD3/KDM6B, and JMJD2C/KDM4C, respectively), presumably via catalytic site iron interaction, displaying little toward 5mC DNA hydroxylase TET1, aurora kinases, HDACs, HIF prolyl hydroxylase PHD2, histone methyltransferases, protein arginine methyltransferases, sirtuins, or the non-jumonji family histone demethylase LSD1/KDM1A. Shown to exhibit anticancer activity against H358 and A549 lunger cancer cells both in cultures in vitro (IC50 in 4 d = 100 and 250 nM, respectively) and in mice in vivo (via i.p. at 100mg/kg and p.o. at 55mg/kg, respectively; 2-3 times per week).

Specifications
Specifications
Color | Light Beige |
Description | 98% by HPLC |
Target Proteins | Jumonji family of histone demethylases |
CAS | 199596-05-9 |
Molecular Formula | C17H13ClN4 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction