Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ KN-93

Catalog No. p-7060094 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

A cell-permeable, reversible and competitive inhibitor of rat brain CaM kinase II (Ki = 370 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins cam kinase 2
CAS 139298-40-1
Molecular Formula C26H29CIN2O4S
Formula Weight 501g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.