Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Roscovitine
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
Inhibits p34cdk1/cyclin B (IC50 = 650 nM), p33cdk2/cyclin A (IC50 = 700 nM), p33cdk2/cyclin E (IC50 = 700 nM), and p33cdk5/p35 (IC50 = 200 nM) by competing for the ATP binding domain of the kinases. Exhibits reduced sensitivity towards related kinases including ERK1 and ERK2 (IC50 = 34μM and 14μM, respectively). Roscovitine does not significantly affect the activity of other protein kinases even at 100μM, including p34cdk4/cyclin D1 and p40cdk6/cyclin D2. Compared to olomoucine, roscovitine displays increased anti-mitotic activity at the G1/S and G2/M phases of the cell cycle. A 50mm (5mg/282μl) solution of Roscovitine (Cat. No. 557364) in DMSO is also available.
Specifications
Specifications
| Color | White to Off-white |
| Target Proteins | p34cdk1/cyclin B |
| CAS | 186692-46-6 |
| Molecular Formula | C19H26N6O |
| Formula Weight | 354.5g/mol |
| Solubility | DMSO |
| Form | Solid |
| Quantity | 5 mg |
| Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction