Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ L-701,324
Click to view available options
:
10mg
Specifications
Specifications
Color | White |
Description | 98% by HPLC |
Target Proteins | NMDA receptors |
Molecular Formula | C21H14CINO3 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Powder |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction