Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LCAT Activity Assay Kit (Fluorometric)

Catalog No. 4289001kit
Click to view available options
Quantity:
1 Kit

Useful for quantitative measurement of lecithin, cholesterol acyltransferase activity.

EMD Millipore Calbiochem™ LCAT Activity Assay Kit (Fluorometric) mediates formation of cholesteryl esters in human plasma.

  • Transfers acyl chain from sn-2 position of phosphatidylcholine to cholesterol
  • Packaging: Glass Bottle

Recommended Storage

Multiple storage requirements

TRUSTED_SUSTAINABILITY

Specifications

Description LCAT Activity Assay Kit (Fluorometric)
Kit Contents LCAT Substrate, READ Reagent, and user protocol
Quantity 1 Kit
Sufficient For 100 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.