Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Caspase Inhibitor III
Shop All MilliporeSigma Products
Click to view available options
Quantity:
250 μg
Description
- Peptide Sequence: Boc-Asp(OMe)-CH2F*
- Cell Permeable

Specifications
Specifications
Color | Off-white |
Target Proteins | broad spectrum caspase |
CAS | 634911-80-1 |
Molecular Formula | C11H18FNO5 |
Formula Weight | 263.3g/mol |
Solubility | DMSO |
Content And Storage | Protect from moisture |
Form | Semi-solid |
Quantity | 250 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction