EMD Millipore Calbiochem™ LY 294002

LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.

Manufacturer: EMD Millipore 4402025MG

 View more versions of this product

Catalog No. 44-020-25MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins Phosphatidylinositol 3-kinase
    Molecular Formula C19H17NO3
    Formula Weight 307.4g/mol
    Solubility DMSO or EtOH
    Description 98% by HPLC
    Format Off-white solid
    Quantity 5mg
    CAS 154447-36-6

    Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit