Learn More
MilliporeSigma™ Calbiochem™ β-Mercaptoethanol, Molecular Biology Grade
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| CAS | 60-24-2 |
| Content And Storage | Store at +15°C to +30°C |
| Formula Weight | 78.1g/mol |
| Molecular Formula | C2H6OS |
| Purity | ≥99% by GC |
| Quantity | 250 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.