Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Milrinone
Click to view available options
Quantity:
10 mg
Description
Cell Permeable

Specifications
Specifications
Color | White |
Description | 99% by HPLC |
Target Proteins | PDE 3 |
CAS | 78415-72-2 |
Molecular Formula | C12H9N3O |
Solubility | DMSO or EtOH |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction