Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MitoBloCK-6

Catalog No. 5057590001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-575-90001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-575-90001 Supplier MilliporeSigma™ Supplier No. 5057590001
Add to Cart
Add to Cart

A cell-permeable, stable dichlorosalicylaldehyde Schiff's base that acts as a potent, selective inhibitor of Mia40/Erv1 redox-mediated import pathway (IC50 = 700 nM, 900 nM, an

A cell-permeable, stable dichlorosalicylaldehyde Schiff′s base that acts as a potent, selective inhibitor of Mia40/Erv1 redox-mediated import pathway (IC50 = 700 nM, 900 nM, and 1.4μM for ALR, Erv1, and Erv2, respectively). Significantly reduces the import of CX9 proteins, Erv1, Tim23, and ADP/ATP carrier (AAC). However, it does not affect mitochondrial membrane integrity as evidenced by the lack of aconitase, AAC, Tim54, Mia40, and cytochrome c release. Has no effect on protein disulfide isomerase, flavin adenine dinucleotide, and succinate dehydrogenase activities and does not disrupt mitochondrial net work or reduce viability of cells even at high concentrations (∽100μM in HeLa and HEK293 cells). Reported to specifically cause cytochrome c release, activate caspase-3, and induce apoptosis in human embryonic stem cells (∽20μM), but not in differentiated cells. Reversibly impairs cardiac development and reduces heart rate in zebra fish that is attributed to mitochondrial dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 98% by HPLC
Target Proteins Erv1
Molecular Formula C19H14Cl2N2O
Formula Weight 357.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.