Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Mitosis Inhibitor, AK301
Click to view available options
:
10mg
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach

Specifications
Specifications
Color | White |
Description | 98% by HPLC |
Target Proteins | tubulin |
Molecular Formula | C19H21ClN2O2 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction