Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MMP-2/MMP-9 Inhibitor III
Click to view available options
:
1mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: H-Cys1-Thr-Thr-His-Trp-Gly-Phe-Thr-Leu-Cys10-OH (cyclic: 1→ 10)

Specifications
Specifications
Color | White |
Formulation | Supplied as a trifluoroacetate salt. |
Description | 97% by HPLC |
Target Proteins | MMP-2, MMP-9 |
Molecular Formula | C52H71N13O14S2 |
Formula Weight | 1166.4g/mol |
Solubility | H2O |
Content And Storage | Hygroscopic |
Form | Lyophilized |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction