Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MNK1 Inhibitor
Click to view available options
:
5mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | Off-white |
Description | 98% by HPLC |
Target Proteins | Mitogen-activated protein kinase-interacting kinase 1 (MNK1) |
CAS | 522629-08-9 |
Molecular Formula | C11H9FN6 |
Formula Weight | 244.2g/mol |
Solubility | DMSO or EtOH |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction