Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MRS 2211
Click to view available options
Quantity:
10 mg
Description
A highly selective and competitive antagonist of P2Y13 purinergic receptor (pIC50 = 5.97). Exhibits about 20-fold greater selectivity over P2Y1 and P2Y12 receptors.

Specifications
Specifications
Color | Dark Red |
Description | 95% by HPLC |
Target Proteins | P2Y13 |
Molecular Formula | C14H9ClN4Na3O8P |
Solubility | H2O |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction