Learn More
MilliporeSigma™ Calbiochem™ NADH, Disodium Salt
Catalog No. 481913500MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
48-191-3500MG 500 mg
1 options
Catalog No. 48-191-3500MG Supplier MilliporeSigma™ Supplier No. 481913500MG
Only null left
Add to Cart
Add to Cart
Functions as a coenzyme for large number of oxidoreductases
Unstable in solution; reconstitute just prior to use.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Additive NADH, Disodium Salt
Quantity 500 mg