Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ NADP, Disodium Salt
Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 mg
1 g
Description
- Coenzyme that is widely distributed in living matter
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Additive | NADP, Disodium Salt |
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction