Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nitric Oxide Synthase Assay Kit (Colorimetric)

Catalog No. 4827021KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-270-21KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-270-21KIT Supplier MilliporeSigma™ Supplier No. 4827021KIT
Add to Cart
Add to Cart

Includes

Assay Buffer, Nitrate Reductase, Cofactors, Nitrate Standard, Lactate Dehydrogenase, Griess Reagents (R1 and R2), Microtiter Plate, Plate Cover, and user protocol

Used for measurement of nitric oxide synthase activity.

EMD Millipore Calbiochem™ Nitric Oxide Synthase Assay Kit (Colorimetric) uses lactate dehydrogenase (LDH) to destroy excess NADPH.

  • Colorimetric detection
  • Format: 96 well plate
  • TotalAssayTime: 2 hr
  • Sufficient for 96 Tests
  • Sample Type: Plasma, serum, urine, tissue culture medium, tissue homogenates
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Quantity 1 Kit
Content And Storage −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.