Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Oligomycin
Click to view available options
:
10mg
Description
A mixture of A, B, and C isomers. A macrolide antibiotic that inhibits membrane-bound mitochondrial ATP synthase, preventing phosphoryl group transfer. Induces apoptosis in cultured human lymphoblastoid and other mammalian cells.

Specifications
Specifications
Color | White |
Description | 90% by HPLC (mixture of A, B, and C isomers) |
Target Proteins | Membrane-bound mitochondrial ATPase (F1) |
CAS | 1404-19-9 |
Molecular Formula | C45H74O11 |
Formula Weight | 791.1g/mol |
Solubility | EtOH |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction