Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Oncrasin-1
Click to view available options
:
25mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | Off-white |
Description | 95% by HPLC |
CAS | 75629-57-1 |
Molecular Formula | C16H12ClNO |
Formula Weight | 269.7g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 25 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction