Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Paclitaxel, 6α-Hydroxy-
Click to view available options
Quantity:
20 nmol
Description
A metabolite of the anticancer agent Paclitaxel (Cat. No. 580555).

Specifications
Specifications
Color | Colorless |
Description | 98% by HPLC |
Target Proteins | A metabolite of the anticancer agent Paclitaxel |
CAS | 153212-75-0 |
Molecular Formula | C47H51NO15 |
Formula Weight | 869.9g/mol |
Solubility | DMSO |
Form | Solid |
Quantity | 20 nmol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction