Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Pepstatin A, Synthetic
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Description
A reversible inhibitor of aspartic proteases. Inhibits cathepsin D, pepsin, and renin. Reported to block apoptosis induced by CA-074 (Cat. No. 205530) in PC12 cell lines. Also available as a 25mm solution in DMSO (Cat. No. 508437).

Specifications
Specifications
Color | White |
Target Proteins | cathepsin D |
CAS | 26305-03-3 |
Molecular Formula | C34H63N5O9 |
Formula Weight | 685.9g/mol |
Solubility | DMSO or MeOH |
Form | Solid |
Quantity | 100 mg |
Purity | 98% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction