Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Phenylmethylsulfonyl Fluoride

Catalog No. 523325GM Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. 52-332-5GM

Supplier: MilliporeSigma™ 523325GM

Only null left
Add to Cart
Add to Cart

Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.

Irreversible inhibitor of serine proteases. Its mechanism of action is analogous to that of diisopropylfluorophosphate. PMSF causes sulfonylation of the active-site serine residues. Also reported to inhibit internucleosomal DNA fragmentation in immature thymocytes. For a related, more stable inhibitor, see AEBSF (Cat. No. 101500).
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Target Proteins serine proteases
CAS 329-98-6
Molecular Formula C7H7FO2S
Formula Weight 174.2g/mol
Solubility EtOH, isopropanol, or MeOH
Content And Storage Protect from moisture
Form Crystalline Solid
Quantity 5 g
Purity 99% by GC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.