Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PI 3-Kα Inhibitor VIII
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Pale Yellow |
| Description | 95% by HPLC |
| CAS | 372196-77-5 |
| Molecular Formula | C16H14N5O4SBr·HCl·2H2O |
| Formula Weight | 524.8g/mol |
| Solubility | DMSO |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Solid |
| Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction