Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PIM-Kinase Inhibitor IX, SGI-1776
Click to view available options
Quantity:
10 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | Off-white |
Description | 99% by HPLC |
Molecular Formula | C20H22F3N5O·2H2SO4 |
Formula Weight | 601.6g/mol |
Solubility | DMSO or H2O |
Content And Storage | Hygroscopic, Protect from light |
Form | Powder |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction