Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PKC Inhibitor Peptide 19-31
Click to view available options
Quantity:
1 mg
Description
More potent, reversible, and substrate competitive inhibitor of PKC (IC50 = 100nM) than PKC Inhibitor 19-36 (Cat. No. 539560). Inhibits both autophosphorylation and protein substrate phosphorylation.

Specifications
Specifications
Color | White to Off-white |
Description | ≥97% by HPLC |
Target Proteins | PKC |
Molecular Formula | C67H118N26O16 |
Formula Weight | 1543.8g/mol |
Solubility | 5% acetic acid |
Content And Storage | Toxicity: Standard Handling, Store at -20°C, Protect from moisture, Hygroscopic, Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. |
Form | Lyophilized |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction