Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PKR Inhibitor III, 7DG CAS 26927-01-5
Click to view available options
Quantity:
5 mg
Description
A cell-permeable gedunin analog that directly targets PKR via reversible interaction inhibits PKR-mediated cellular events by disrupting PKR signaling complexes assembly, such as IκK complex and inflammasome/pyroptosome, resulting in effective blockage of LPS-induced IκBβ degradaion (20μM) and protection against anthrax toxin LT-induced caspase-1 activation as well as subsequent pyroptosis (10μM) in murine macrophage J774 cultures. PKR is also known to mediate LT-induced apoptosis via its kinase activity in LPS-primed Macrophages derived from C57BL/6 with a resistent NLRP1 allele, inhibitable by C16 (Cat. Nos. 527450 and 527451), but not by 7DG, while LT-induced pyroptosis in J774 cultures is insensitive to C16 treatment.

Specifications
Specifications
Color | Pale Yellow |
Description | 95% by HPLC |
Target Proteins | PKR |
Molecular Formula | C26H30O5 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction