Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PP1 Analog
Click to view available options
Quantity:
1 mg
Description
A potent, reversible, ATP-competitive, and cell-permeable inhibitor of Src-family tyrosine kinases that is > 800 times more selective for the I338G mutant v-Src (IC50 = 1.5nM) compared to wild-type v-Src (IC50 = 1.0μM). Also inhibits wild-type Fyn (IC50 = 600nM). Also available in InSolution™ format (Cat. No. 529605).

Specifications
Specifications
Color | Off-white |
Description | 99% by HPLC |
Target Proteins | I338G mutant v-Src |
CAS | 221243-82-9 |
Molecular Formula | C19H19N5 |
Formula Weight | 317.4g/mol |
Solubility | DMSO |
Form | Solid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction