Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PPM1D Phosphatase Inhibitor II, GSK2830371
Click to view available options
Quantity:
10 mg
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach

Specifications
Specifications
Color | Light Yellow |
Description | 97% by HPLC |
Target Proteins | Wip1/PPM1D/PP2Cδ |
Molecular Formula | C23H29ClN4O2S |
Solubility | DMSO |
Form | Powder |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction