Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set I, Animal-Free

Catalog No. 5351421SET
Click to view available options
Quantity:
1 Set
1 mL

The Protease Inhibitor Cocktail Set I, Animal-Free controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A cocktail of five protease inhibitors for the inhibition of a broad range of proteases and esterases. Each vial, when reconstituted with 1mL H2O, yields a 100X stock solution. When diluted to 1X the cocktail contains 500μM AEBSF, HCl (Cat. No. 101500), 150nM Aprotinin (Cat. No.616371), 1μM E-64 (Cat. No. 324890), 0.5mm EDTA, Disodium Salt and 1μM Leupeptin Hemisulfate (Cat. No.108975). Available as 1mL or as a set of 10 x 1 ml.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins A broad range of proteases and esterases
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 Set
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.