Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Proteasome Inhibitor II
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
- Peptide Sequence: Z-Leu-Leu-Phe-CHO
- Cell Permeable

Specifications
Specifications
Color | White |
Target Proteins | MPC |
Molecular Formula | C29H39N3O5 |
Formula Weight | 509.7g/mol |
Solubility | DMSO |
Content And Storage | Protect from moisture |
Form | Lyophilized |
Quantity | 5 mg |
Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction