Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Rac1 Inhibitor
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
10 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Target Proteins | Rac1 |
CAS | 1177865-17-6 |
Molecular Formula | C24H38Cl3N7 |
Formula Weight | 530.96g/mol |
Solubility | H2O |
Content And Storage | Hygroscopic, Protect from light |
Form | Lyophilized |
Quantity | 5 mg |
Purity | 93% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction