Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Rb/Raf-1 Disruptor 251
Click to view available options
Quantity:
25 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | White |
Description | 97% by HPLC |
CAS | 72214-67-6 |
Molecular Formula | C8H9Cl3N2S |
Formula Weight | 271.6g/mol |
Solubility | DMSO or EtOH |
Content And Storage | Hygroscopic, Protect from light |
Form | Solid |
Quantity | 25 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction