Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 202190

Catalog No. 5593881mg Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg

SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (K

A potent, reversible, competitive, and cell-permeable inhibitor of p38 MAP kinase. Inhibits p38 phosphorylation of myelin basic protein (MBP) with no effect on the activity of the ERK or JNK MAP kinase subgroups. Also inhibits the kinase activity of p38β (Ki = 16nM; IC50 = 350nM) and p38 phosphorylation of activating transcription factor 2 (ATF-2; IC50 = 280nM). Blocks LPS-induced TNF-α and interleukin biosynthesis. Reported to induce LDL receptor expression in Hep52 cells. A 1mg/mL solution of SB 202190 (Cat. No. 559397) in anhydrous DMSO is also available.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins p38β
CAS 152121-30-7
Molecular Formula C20H14FN3O
Formula Weight 331.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.