Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 203580

Catalog No. 5593895MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg

SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins P38MAPK
CAS 152121-47-6
Molecular Formula C21H16N3OSF
Formula Weight 377.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Thin Film
Quantity 5 mg
Purity 98% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.