Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ SB 203580
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Description
Cell Permeable
Specifications
Specifications
| Target Proteins | P38MAPK |
| CAS | 152121-47-6 |
| Molecular Formula | C21H16N3OSF |
| Formula Weight | 377.4g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Thin Film |
| Quantity | 10 mg |
| Purity | 98% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction