Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT3 Inhibitor Peptide, Cell-Permeable

Catalog No. 5730965MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

The STAT3 Inhibitor Peptide, Cell-Permeable controls the biological activity of STAT3.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Pro-Tyr-(PO3H2)-Leu-Lys-Thr-Lys-Ala-Ala-Val-Leu-Leu-Pro-Val-Leu-Leu-Ala-Ala-Pro-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Target Proteins Blocker of Stat3 activation
Molecular Formula C92H157N20O24P
Formula Weight 1958.4g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 5 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.