Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Nα-Tosyl-Lys Chloromethyl Ketone, Hydrochloride
Shop All MilliporeSigma Products
Click to view available options
Quantity:
50 mg
250 mg
Description
Inhibits trypsin-like serine proteinases. Irreversibly inactivates trypsin without affecting chymotrypsin. Prevents nitric oxide production by activated macrophages by interfering with transcription of the iNOS gene. Blocks cell-cell adhesion and binding of HIV-1 virus to the target cells. In macrophages, blocks nitric oxide synthase induced by interferon-γ and lipopolysaccharides (EC50 = 80μM). Prevents endonucleolysis accompanying apoptotic death of HL-60 leukemia cells and normal thymocytes.
Specifications
Specifications
| Color | Off-white |
| Target Proteins | trypsin like proteases |
| CAS | 4238-41-9 |
| Molecular Formula | C14H21ClN2O3S·HCl |
| Formula Weight | 369.3g/mol |
| Solubility | Aqueous solutions (unstable above pH 6.0) |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Quantity | 50 mg |
| Purity | 95% by elemental analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction