Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Wiskostatin

Catalog No. p-7060181 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

A cell-permeable N-alkylated carbazole derivative that selectively blocks actin filament assembly.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins N-WASP (neural Wiskott Aldrich syndrome protein)
CAS 253449-04-6
Molecular Formula C17H18Br2N2O
Formula Weight 426.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.