Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ ADP-HPD, Dihydrate, Ammonium Salt
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Set
60 μg
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | White |
| Target Proteins | PARG |
| Molecular Formula | C15H23N6O12P2·NH4·2H2O |
| Formula Weight | 595.4g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Lyophilized |
| Quantity | 1 Set |
| Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction