Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TCEP, Hydrochloride

Catalog No. 5805601gm
Click to view available options
Quantity:
1 g

Reduces even the most stable water-soluble alkyl disulfides at pH 4.5 within 5 minutes

  • Water-soluble and odorless disulfide-reducing agent that is more stable and effective than DTT
  • Also capable of functioning as a reducing agent even in the pH range of 1.5-8.5
  • It is more resistant to air-oxidation as compared to DTT
TRUSTED_SUSTAINABILITY

Specifications

Purity ≥98% by elemental analysis
CAS 51805-45-9
Content And Storage 2°C to 8°C
Form Crystalline Solid
Quantity 1 g
Synonym tris(2-Carboxyethyl)phosphine, HCl
Solubility Water
Product Type Calbiochem TCEP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.