Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Ubiquitinated Protein Enrichment Kit
Click to view available options
Quantity:
1 Kit
Description
EMD Millipore Calbiochem™ Ubiquitinated Protein Enrichment Kit is rapid method useful for enrichment of polyubiquitinated proteins from cell tissue lysates.
- Contains ubiquitin-associated sequence bound to glutathione-agarose
- Useful for enrichment of polyubiquitinated proteins from cell tissue lysates of broad range of species including canine, human, mouse, yeast
- Can be identified by loading beads directly onto SDS-PAGE, immunoblotting with antibody of choice or Anti-Ubiquitin
- Packaging: Glass Bottle
Recommended Storage
2° to 8°C

Specifications
Specifications
Description | Ubiquitinated Protein Enrichment Kit |
Kit Contents | Polyubiquitin Affinity Beads, Control Glutathione-Agarose Beads, Control Lysate, and user protocol |
Quantity | 1 Kit |
Sufficient For | To process 12.5 to 25mg lysate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction